missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myelin protein P0 Protein

A DNA sequence encoding the Homo sapiens (Human) Myelin protein P0, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8589-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055441

  • 1210.00€ / 500 microgramos


Fecha estimada de envío
13-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000521.2
Myelin protein P0 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
4359
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MPZ
Recombinant Protein
E. coli
IVVYTDREVH GAVGSRVTLH CSFWSSEWVS DDISFTWRYQ PEGGRDAISI FHYAKGQPYI DEVGTFKERI QWVGDPRWKD GSIVIHNLDY SDNGTFTCDV KNPPDIVGKT SQVTLYVFEK VPTRYGV
Videos
Documentos