missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MXD3 Protein

A cDNA sequence encoding the MXD3 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12764-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15931469

  • 3185.00€ / 1 miligramo


Fecha estimada de envío
30-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

MXD3 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MXD3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMEPLASN IQVLLQAAEF LERREREAEH GYASLCPHRS PGPIHRRKKR PPQAPGAQDS GRSVHNELEK RRRAQLKRCL ERLKQQMPLG ADCARYTTLS LLRRARMHIQ KLEDQEQRAR QLKERLRSKQ QSLQRQLEQL RGLAGAAERE RLRADSLDSS GLSSERSDSD QEELEVDVES LVFGGEAELL RGFVAGQEHS YSHGGGAWL
Videos
Documentos