missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MX2 Protein

A cDNA sequence encoding the MX2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12763-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15911469

  • 160.00€ / 20 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

MX2 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MX2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSIGIH LNAYFLETSK RLANQIPFII QYFMLRENGD SLQKAMMQIL QEKNRYSWLL QEQSETATKR RILKERIYRL TQARHALCQF SSKEIH
Videos