missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Muscleblind-like protein 1

A DNA sequence encoding the Homo sapiens (Human) Muscleblind-like protein 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8222-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053371

  • 160.00€ / 10 microgramos


Fecha estimada de envío
21-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001300986.1
Muscleblind-like protein 1
10 μg
Not determined.
Full Length of Isoform 5
Human
His-SUMO
Tris-based buffer, 50% glycerol
4154
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MBNL1
Recombinant Protein
E. coli
MAVSVTPIRD TKWLTLEVCR EFQRGTCSRP DTECKFAHPS KSCQVENGRV IACFDSLKGR CSRENCKYLH PPPHLKTQLE INGRNNLIQQ KNMAMLAQQM QLANAMMPGA PLQPVPMFSV APSLATNASA AAFNPYLGPV SPSLVPAEIL PTAPMLVTGN PGVPVPAAAA AAAQKLMRTD RLEVCREYQR GNCNRGENDC RFAHPADSTM IDTNDNTVTV CMDYIKGRCS REKCKYFHPP AHLQAKIKAA QYQVNQAAAA QAAATAAAMG IPQAVLPPLP KRPALEKTNG ATAVFNTGIF QYQQALANMQ LQQHTAFLPP GSILCMTPAT SVVPMVHGAT PATVSAATTS ATSVPFAATA TANQIPIISA EHLTSHKYVT QM
Videos
Documentos