missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MTPN Protein
A cDNA sequence encoding the MTPN was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12750-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MTPN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MCDKEFMWAL KNGDLDEVKD YVAKGEDVNR TLEGGRKPLH YAADCGQLEI LEFLLLKGAD INAPDKHHIT PLLSAVYEGH VSCVKLLLSK GADKTVKGPD GLTAFEATDN QAIKALLQ | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MTPN Human solution containing 20mM Tris-HCl pH 8 and 10% glycerol. |