missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MTHFD2 Protein
A cDNA sequence encoding the MTHFD2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12749-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
10797 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLAAVRNE AVVISGRKLA QQIKQEVRQE VEEWVASGNK RPHLSVILVG ENPASHSYVL NKTRAAAVVG INSETIMKPA SISEEELLNL INKLNNDDNV DGLLVQLPLP EHIDERRICN AVSPDKDVDG FHVINVGRMC LDQYSMLPAT PWGVWEIIKR TGIPTLGKNV VVAGRSKNVG MPIAMLLHTD GAHERPGGDA TVTISHRYTP KEQLKKHTIL ADIVISAAGI PNLITADMIK EGAAVIDVGI NRVHDPVTAK PKLVGDVDFE GVRQKAGYIT PVPGGVGPMT VAMLMKNTII AAKKVLRLEE REVLKSKELG VATN | |
Greater than 95.0% as determined by SDS-PAGE. |
MTHFD2 Protein | |
Research Use Only | |
MTHFD2 | |
Human | |
His | |
The MTHFD2 solution (1 mg/ml) contains Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |