missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MORF4L1 Protein
A cDNA sequence encoding the MORF4L1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12719-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MORF4L1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAPKQD PKPKFQEGER VLCFHGPLLY EAKCVKVAIK DKQVKYFIHY SGWNKNWDEW VPESRVLKYV DTNLQKQREL QKANQEQYAE GKMRGAAPGK KTSGLQQKNV EVKTKKNKQK TPGNGDGGST SETPQPPRKK RARVDPTVEN EETFMNRVEV KVKIPEELKP WLVDDWDLIT RQKQLFYLPA KKNVDSILED YANYKKSRGN TDNKEYAVNE VVAGIKEYFN VMLGTQLLYK FERPQYAEIL ADHPDAPMSQ VYGAPHLLRL FVRIGAMLAY TPLDEKSLAL LLNYLHDFLK YLAKNSATLF SASDYEVAPP EYHRKAV | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MORF4L1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.15M NaCl and 1mM DTT. |