missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MORC family CW-type zinc finger protein 1
A DNA sequence encoding the Homo sapiens (Human) MORC family CW-type zinc finger protein 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7595-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_056173.1 | |
MORC family CW-type zinc finger protein 1 | |
Research Use Only | |
MORC3 | |
Recombinant Protein | |
E. coli | |
MAAQPPRGIR LSALCPKFLH TNSTSHTWPF SAVAELIDNA YDPDVNAKQI WIDKTVINDH ICLTFTDNGN GMTSDKLHKM LSFGFSDKVT MNGHVPVGLY GNGFKSGSMR LGKDAIVFTK NGESMSVGLL SQTYLEVIKA EHVVVPIVAF NKHRQMINLA ESKASLAAIL EHSLFSTEQK LLAELDAIIG KKGTRIIIWN LRSYKNATEF DFEKDKYDIR IPEDLDEITG KKGYKKQERM DQIAPESDYS LRAYCSILYL KPRMQIILRG QKVKTQLVSK SLAYIERDVY | |
Greater than 90% as determined by SDS-PAGE. |
23515 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |