missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMP 8 Protein
A cDNA sequence encoding the MMP 8 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10789-10U
Additional Details : Peso : 0.01000kg
Specifications
4317 | |
10 IU | |
< 1.0 EU per ug protein as determined by the LAL method. | |
100 units/ml after activation with APMA by solution assay method. One unit of collagenolytic activity is defined as the cleavage of 1microg of collagen per minute by the solution method. | |
Human | |
Untagged | |
The MMP-8 protein solution (100 units/ml) in 0.05M Tris-HCl buffer, pH 7.6, 0.2M NaCl, 5mM CaCl2, 0.0025% NaN3 and 0.1% BSA. |
MMP 8 Protein | |
Research Use Only | |
MMP8 | |
Recombinant Protein | |
E. coli | |
MFSLKTLPFL LLLHVQISKA FPVSSKEKNT KTVQDYLEKF YQLPSNQYQS TRKNGTNVIV EKLKEMQRFF GLNVTGKPNE ETLDMMKKPR CGVPDSGGFM LTPGNPKWER TNLTYRIRNY TPQLSEAEVE RAIKDAFELW SVASPLIFTR ISQGEADINI AFYQRDHGDN SPFDGPNGIL AHAFQPGQGI GGDAHFDAEE TWTNTSANYN LFLVAAHEFG HSLGLAHSSD PGALMYPNYA FRETSNYSLP QDDIDGIQAI YGLSSNPIQP TGPSTPKPCD PSLTFDAITT LRGEILFFKD RYFWRRHPQL QRVEMNFISL FWPSLPTGIQ AAYEDFDRDL IFLFKGNQYW ALSGYDILQG YPKDISNYGF PSSVQAIDAA VFYRSKTYFF VNDQFWRYDN QRQFMEPGYP KSISGAFPGI ESKVDAVFQQ EHFFHVFSGP RYYAFDLIAQ RVTRVARGNK WLNCRYG | |
Greater than 90% as determined by SDS-PAGE. |