missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MMP-3 Protein

A DNA sequence encoding the Homo sapiens (Human) MMP-3 Protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6370-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052810

  • 103.00€ / 10 microgramos


Fecha estimada de envío
21-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_002413.1
MMP-3 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
4314
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MMP3
Recombinant Protein
E. coli
TFPGIPKWRK THLTYRIVNY TPDLPKDAVD SAVEKALKVW EEVTPLTFSR LYEGEADIMI SFAVREHGDF YPFDGPGNVL AHAYAPGPGI NGDAHFDDDE QWTKDTTGTN LFLVAAHEIG HSLGLFHSAN TEALMYPLYH SLTDLTRFRL SQDDINGIQS LYGPPPDSPE TPLVPTEPVP PEPGTPANCD PALSFDAVST LRGEILIFKD RHFWRKSLRK LEPELHLISS FWPSLPSGVD AAYEVTSKDL VFIFKGNQFW AIRGNEVRAG YPRGIHTLGF PPTVRKIDAA ISDKEKNKTY FFVEDKYWRF DEKRNSMEPG FPKQIAEDFP GIDSKIDAVF EEFGFFYFFT GSSQLEFDPN AKKVTHTLKS NSWLNC
Videos
Documentos