missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MMAB Protein

A cDNA sequence encoding the MMAB was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12702-25ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15921279

Cantidad Precio
1 161.00€ / 25 microgramos
10 150.00€ / 25 microgramos
Fecha estimada de envío
06-08-2020
Añadir a la cesta


Especificaciones

Especificaciones

MMAB Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MMAB 1 mg/ml protein solution contains 20mM Tris pH 7.5 and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQSRGPQGVE DGDRPQPSSK TPRIPKIYTK TGDKGFSSTF TGERRPKDDQ VFEAVGTTDE LSSAIGFALE LVTEKGHTFA EELQKIQCTL QDVGSALATP CSSAREAHLK YTTFKAGPIL ELEQWIDKYT SQLPPLTAFI LPSGGKISSA LHFCRAVCRR AERRVVPLVQ MGETDANVAK FLNRLSDYLF TLARYAAMKE GNQEKIYKKN DPSAESEGL
Videos
Documentos