missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MKI67 Protein

A DNA sequence encoding the Homo sapiens (Human) MKI67, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6361-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052750

  • 103.00€ / 10 microgramos


Fecha estimada de envío
23-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001139438.1
MKI67 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
4288
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MKI67
Recombinant Protein
Yeast
NEKKPMKTSP EMDIQNPDDG ARKPIPRDKV TENKRCLRSA RQNESSQPKV AEESGGQKSA KVLMQNQKGK GEAGNSDSMC LRSRKTKSQP AASTLESKSV QRVTRSVKRC AENPKKAEDN VCVKKIRTRS HRDSEDI
Videos
Documentos