missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mitochondrial fission process protein 1

A DNA sequence encoding the Homo sapiens (Human) Mitochondrial fission process protein 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8083-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052561

  • 245.00€ / 10 microgramos


Fecha estimada de envío
25-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001003704.1
Mitochondrial fission process protein 1
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
51537
Greater than 80% as determined by SDS-PAGE.
Research Use Only
MTFP1
Recombinant Protein
E. coli
MSEPQPRGAE RDLYRDTWVR YLGYANEVGE AFRSLVPAAV VWLSYGVASS YVLADAIDKG KKAGEVPSPE AGRSARVTVA VVDTFVWQAL ASVAIPGFTI NRVCAASLYV LGTATRWPLA VRKWTTTALG LLTIPIIIHP IDRSVDFLLD SSLRKLYPTV GKPSSS
Videos
Documentos