missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mitochondrial fission process protein 1
A DNA sequence encoding the Homo sapiens (Human) Mitochondrial fission process protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8083-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001003704.1 | |
Mitochondrial fission process protein 1 | |
Research Use Only | |
MTFP1 | |
Recombinant Protein | |
E. coli | |
MSEPQPRGAE RDLYRDTWVR YLGYANEVGE AFRSLVPAAV VWLSYGVASS YVLADAIDKG KKAGEVPSPE AGRSARVTVA VVDTFVWQAL ASVAIPGFTI NRVCAASLYV LGTATRWPLA VRKWTTTALG LLTIPIIIHP IDRSVDFLLD SSLRKLYPTV GKPSSS | |
Greater than 80% as determined by SDS-PAGE. |
51537 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |