Learn More
enQuireBio™ Recombinant Human MIEN1 Protein
A cDNA sequence encoding the MIEN1 was constructed and used to recombinantly synthesize the protein.
Marca:  enQuireBio™ QP12695-5ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15921259
  • 137.00€ / 5 microgramos
Fecha estimada de envío: 28-05-2024
para ver el stock
Especificaciones
MIEN1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSGEPGQ TSVAPPPEEV EPGSGVRIVV EYCEPCGFEA TYLELASAVK EQYPGIEIES RLGGTGAFEI EINGQLVFSK LENGGFPYEK DLIEAIRRAS NGETLEKITN SRPPC
Greater than 95.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MIEN1 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.