missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Midkine / MDK Protein

A DNA sequence encoding the Human Midkine / MDK Protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10419-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933798

  • 1095.00€ / 100 microgramos


Fecha estimada de envío
26-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001012333.1
Midkine / MDK Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
VAKKKDKVKK GGPGSECAEW AWGPCTPSSK DCGVGFREGT CGAQTQRIRC RVPCNWKKEF GADCKYKFEN WGACDGGTGT KVRQGTLKKA RYNAQCQETI RVTKPCTPKT KAKAKAKKGK GKD
4192
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
MDK
The biological activity determined by a chemotaxis bioassay using human neutrophils is in a concentration range of 0.1-10 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH7.4, lyophilized
Videos
Documentos