Learn More
enQuireBio™ Recombinant Human Microtubule-associated protein 1S Protein
A DNA sequence encoding the Homo sapiens (Human) Microtubule-associated protein 1S, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8768-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001295292.1 | |
Microtubule-associated protein 1S Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55201 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
MAP1S | |
Recombinant Protein | |
E. coli | |
LSDSDPVPLA PGAADSDEDT EGFGVPRHDP LPDPLKVPPP LPDPSSICMV DPEMLPPKTA RQTENVSRTR KPLARPNSRA AAPKATPVAA AKTKGLAGGD RASRPLSARS EPSEKGGRAP LSRKSSTPKT ATRGPSGSAS SRPGVSATPP KSPVYLDLAY LPSGSSAHLV DEEFFQRVRA LCYVISGQDQ RKEEGMRAVL DALLASKQHW DRDLQVTLIP TFDSVAMHTW YAETHARHQA LGITVLGSNS MVSMQDDAFP |