missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MICB Protein

A DNA sequence encoding the Human MICB, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10173-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922968

  • 414.00€ / 100 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001276089.1
MICB Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
AEPHSLRYNL MVLSQDESVQ SGFLAEGHLD GQPFLRYDRQ KRRAKPQGQW AEDVLGAKTW DTETEDLTEN GQDLRRTLTH IKDQKGGLHS LQEIRVCEIH EDSSTRGSRH FYYDGELFLS QNLETQESTV PQSSRAQTLA MNVTNFWKED AMKTKTHYRA MQADCLQKLQ RYLKSGVAIR RTVPPMVNVT CSEVSEGNIT VTCRASSFYP RNITLTWRQD GVSLSHNTQQ WGDVLPDGNG TYQTWVATRI RQGEEQRFTC YMEHSGNHGT HPVPSGKVLV LQSQRTD
4277
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
MICB
The specific activity is determined by binding MICB antibody in ELISA.
Human
Untagged
0.2 um filtered 20 mM Tris, 150 mM NaCl, pH 8.0, lyophilized
Videos