missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MFAP2 Protein

A cDNA sequence encoding the MFAP2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12689-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15911239

  • 103.00€ / 2 microgramos


Fecha estimada de envío
20-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

MFAP2 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MFAP2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1M Urea and 20% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMQGQYDL DPLPPFPDHV QYTHYSDQID NPDYYDYQEV TPRPSEEQFQ FQSQQQVQQE VIPAPTPEPG NAELEPTEPG PLDCREEQYP CTRLYSIHRP CKQCLNEVCF YSLRRVYVIN KEICVRTVCA HEELLRADLC RDKFSKCGVM ASSGLCQSVA ASCARSCGSC
Videos
Documentos