missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Methionine synthase Protein

A DNA sequence encoding the Homo sapiens (Human) Methionine synthase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8028-ye-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052251

  • 805.00€ / 200 microgramos


Fecha estimada de envío
20-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000245.2
Methionine synthase Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
4548
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MTR
Recombinant Protein
Yeast
SLKERRYLPL SQARKSGFQM DWLSEPHPVK PTFIGTQVFE DYDLQKLVDY IDWKPFFDVW QLRGKYPNRG FPKIFNDKTV GGEARKVYDD AHNMLNTLIS QKKLRARGVV GFWPAQSIQD DIHLYAEAAV PQAAEPIATF YGLRQQAEKD SASTEPYYCL SDFIAPLHSG IRDYLGLFAV ACFGVEELSK AYEDDGDDYS SIMVKALGDR LAEAFAEELH ERVRRELWAY CGSEQLDVAD LRRLRYKGIR PAPGYPSQPD HTEKLTMWRL ADIEQSTGIR LTESLAMAPA SAVSGLYFSN LKSKYFAVGK ISKDQVEDYA LRKNISVAEV EKWLGPILGY DTD
Videos
Documentos