missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human METAP2 / MAP2 Protein
A DNA sequence encoding the Homo sapiens (Human) METAP2 / MAP2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6355-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001304111.1 | |
METAP2 / MAP2 Protein | |
Research Use Only | |
METAP2 | |
Recombinant Protein | |
E. coli | |
AGVEEVAASG SHLNGDLDPD DREEGAASTA EEAAKKKRRK KKKSKGPSAA GEQEPDKESG ASVDEVARQL ERSALEDKER DEDDEDGDGD GDGATGKKKK KKKKKRGPKV QTDPPSVPIC DLYPNGVFPK GQECEYPPTQ DGRTAAWRTT SEEKKALDQA SEEIWNDFRE AAEAHRQVRK YVMSWIKPGM TMIEICEKLE DCSRKLIKEN GLNAGLAFPT GCSLNNCAAH YTPNAGDTTV LQYDDICKID FGTHISGRII DCAFTVTFNP KYDTLLKAVK DATNTGIKCA GIDVRLCDVG EAIQEVMESY EVEIDGKTYQ VKPIRNLNGH SIGQYRIHAG KTVPIVKGGE ATRMEEGEVY AIETFGSTGK GVVHDDMECS HYMKNFDVGH VPIRLPRTKH LLNVINENFG TLAFCRRWLD RLGESKYLMA LKNLCDLGIV DPYPPLCDIK GSYTAQFEHT ILLRPTCKEV VSRGDDY | |
Greater than 90% as determined by SDS-PAGE. |
10988 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |