missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Metallothionein-1X Protein

A DNA sequence encoding the Homo sapiens (Human) Metallothionein-1X, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8896-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16057051

  • 1926.00€ / 1 miligramo


Fecha estimada de envío
23-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_005943.1
Metallothionein-1X Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
4501
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MT1X
Recombinant Protein
E. coli
MDPNCSCSPV GSCACAGSCK CKECKCTSCK KSCCSCCPVG CAKCAQGCIC KGTSDKCSC
Videos
Documentos