missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MCP-3 / CCL7 Protein

Recombinant Protein

Marca:  enQuireBio™ QP5253-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915779

  • 1972.00€ / 500 microgramos


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

MCP-3 / CCL7 Protein
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Human MCP-3 / CCL7 induces THP-1 chemotaxis. Activity was further confirmed by measuring primary human neutrophil chemotaxis.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Greater than 95% as determined by SDS-PAGE.
Research Use Only
CCL7
Recombinant Protein
E. coli
QPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKL
Videos