missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAT1A Protein
A cDNA sequence encoding the MAT1A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12649-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MAT1A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHS SGLVPRGSHM NGPVDGLCDH SLSEGVFMFT SESVGEGHPD KICDQISDAV LDAHLKQDPN AKVACETVCK TGMVLLCGEI TSMAMVDYQR VVRDTIKHIG YDDSAKGFDF KTCNVLVALE QQSPDIAQCV HLDRNEEDVG AGDQGLMFGY ATDETEECMP LTIILAHKLN ARMADLRRSG LLPWLRPDSK TQVTVQYMQD NGAVIPVRIH TIVISVQHNE DITLEEMRRA LKEQVIRAVV PAKYLDEDTV YHLQPSGRFV IGGPQGDAGV TGRKIIVDTY GGWGAHGGGA FSGKDYTKVD RSAAYAARWV AKSLVKAGLC RRVLVQVSYA IGVAEPLSIS IFTYGTSQKT ERELLDVVHK NFDLRPGVIV RDLDLKKPIY QKTACYGHFG RSEFPWEVPR KLVF | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MAT1A protein solution contains 20mM Tris-HCl pH 8.0, 1mM DTT, 100mM NaCl and 10% glycerol. |