missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAPK1 Protein
A cDNA sequence encoding the MAPK1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12639-EC-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
5594 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAAAAAAGAG PEMVRGQVFD VGPRYTNLSY IGEGAYGMVC SAYDNVNKVR VAIKKISPFE HQTYCQRTLR EIKILLRFRH ENIIGINDII RAPTIEQMKD VYIVQDLMET DLYKLLKTQH LSNDHICYFL YQILRGLKYI HSANVLHRDL KPSNLLLNTT CDLKICDFGL ARVADPDHDH TGFLTEYVAT RWYRAPEIML NSKGYTKSID IWSVGCILAE MLSNRPIFPG KHYLDQLNHI LGILGSPSQE DLNCIINLKA RNYLLSLPHK NKVPWNRLFP NADSKALDLL DKMLTFNPHK RIEVEQALAH PYLEQYYDPS DEPIAEAPFK FDMELDDLPK EKLKELIFEE TARFQPGYRS | |
Greater than 95.0% as determined by SDS-PAGE. |
MAPK1 Protein | |
Research Use Only | |
MAPK1 | |
Human | |
His | |
MAPK1 is supplied containing 50mM Tris-HCL, 150mM NaCl, 2mM DTT, pH 8.0, 100 Units*/mg. |