missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MANF / ARMET Protein

A DNA sequence encoding the Human MANF / ARMET, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10418-ec-25ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913798

  • 299.00€ / 25 microgramos


Fecha estimada de envío
23-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_006001.4
MANF / ARMET Protein
25 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
LRPGDCEVCI SYLGRFYQDL KDRDVTFSPA TIENELIKFC REARGKENRL CYYIGATDDA ATKIINEVSK PLAHHIPVEK ICEKLKKKDS QICELKYDKQ IDLSTVDLKK LRVKELKKIL DDWGETCKGC AEKSDYIRKI NELMPKYAPK AASARTDL
7873
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
MANF
The ED50 as determined by a cell proliferation assay using rat C6 cells is less than 20 ug/ml, corresponding to a specific activity of > 50 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos