missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mammaglobin-A Protein

A DNA sequence encoding the Homo sapiens (Human) Mammaglobin-A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7590-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16049600

  • 437.00€ / 100 microgramos


Fecha estimada de envío
26-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_002402.1
Mammaglobin-A Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
4250
Greater than 80% as determined by SDS-PAGE.
Research Use Only
SCGB2A2
Recombinant Protein
E. coli
GSGCPLLENV ISKTINPQVS KTEYKELLQE FIDDNATTNA IDELKECFLN QTDETLSNVE VFMQLIYDSS LCDLF
Videos
Documentos