missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LRP4 Protein
A DNA sequence encoding the Homo sapiens (Human) LRP4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6320-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002325.2 | |
LRP4 Protein | |
Research Use Only | |
LRP4 | |
Recombinant Protein | |
Yeast | |
YRHKKSKFTD PGMGNLTYSN PSYRTSTQEV KIEAIPKPAM YNQLCYKKEG GPDHNYTKEK IKIVEGICLL SGDDAEWDDL KQLRSSRGGL LRDHVCMKTD TVSIQASSGS LDDTETEQLL QEEQSECSSV HTAATPERRG SLPDTGWKHE RKLSSESQV | |
Greater than 95% as determined by SDS-PAGE. |
4038 | |
200 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |