missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LRG1 Protein
A cDNA sequence encoding the LRG1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12583-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
LRG1 Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
VTLSPKDCQV FRSDHGSSIS CQPPAEIPGY LPADTVHLAV EFFNLTHLPA NLLQGASKLQ ELHLSSNGLE SLSPEFLRPV PQLRVLDLTR NALTGLPPGL FQASATLDTL VLKENQLEVL EVSWLHGLKA LGHLDLSGNR LRKLPPGLLA NFTLLRTLDL GENQLETLPP DLLRGPLQLE RLHLEGNKLQ VLGKDLLLPQ PDLRYLFLNG NKLARVAAGA FQGLRQLDML DLSNNSLASVPEGLWASLGQ PNWDMRDGFD ISGNPWICDQ NLSDLYRWLQ AQKDKMFSQN DTRCAGPEAV KGQTLLAVAK SQ | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
LRG1 protein filtered (0.4microm) and lyophilized from 20mM Tris and 20mM NaCl, pH 8. |