missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIPA Protein
A DNA sequence encoding the Homo sapiens (Human) LIPA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6311-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000226.2 | |
LIPA Protein | |
Research Use Only | |
LIPA | |
Recombinant Protein | |
Yeast | |
SGGKLTAVDP ETNMNVSEII SYWGFPSEEY LVETEDGYIL CLNRIPHGRK NHSDKGPKPV VFLQHGLLAD SSNWVTNLAN SSLGFILADA GFDVWMGNSR GNTWSRKHKT LSVSQDEFWA FSYDEMAKYD LPASINFILN KTGQEQVYYV GHSQGTTIGF IAFSQIPELA KRIKMFFALG PVASVAFCTS PMAKLGRLPD HLIKDLFGDK EFLPQSAFLK WLGTHVCTHV ILKELCGNLC FLLCGFNERN LNMSRVDVYT THSPAGTSVQ NMLHWSQAVK FQKFQAFDWG SSAKNYFHYN QSYPPTYNVK DMLVPTAVWS GGHDWLADVY DVNILLTQIT NLVFHESIPE WEHLDFIWGL DAPWRLYNKI INLMRKYQ | |
Greater than 90% as determined by SDS-PAGE. |
3988 | |
500 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |