missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LIN28B Protein

A cDNA sequence encoding the LIN28B was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12572-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15920859

Cantidad Precio
1 161.00€ / 5 microgramos
10 150.00€ / 5 microgramos
Fecha estimada de envío
10-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

LIN28B Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The LIN28B solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT and 10% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAEGGAS KGGGEEPGKL PEPAEEESQV LRGTGHCKWF NVRMGFGFIS MINREGSPLD IPVDVFVHQS KLFMEGFRSL KEGEPVEFTF KKSSKGLESI RVTGPGGSPC LGSERRPKGK TLQKRKPKGD RCYNCGGLDH HAKECSLPPQ PKKCHYCQSI MHMVANCPHK NVAQPPASSQ GRQEAESQPC TSTLPREVGG GHGCTSPPFP QEARAEISER SGRSPQEASS TKSSIAPEEQ SKKGPSVQKR KKT
Videos
Documentos