missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIM domain-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) LIM domain-binding protein 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6290-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001073583.1 | |
LIM domain-binding protein 1 | |
Research Use Only | |
LDB3 | |
Recombinant Protein | |
E. coli | |
MSYSVTLTGP GPWGFRLQGG KDFNMPLTIS RITPGSKAAQ SQLSQGDLVV AIDGVNTDTM THLEAQNKIK SASYNLSLTL QKSKRPIPIS TTAPPVQTPL PVIPHQKVVV NSPANADYQE RFNPSALKDS ALSTHKPIEV KGLGGKATII HAQYNTPISM YSQDAIMDAI AGQAQAQGSD FSGSLPIKDL AVDSASPVYQ AVIKSQNKPE DEADEWARRS SNLQSRSFRI LAQMTGTEFM QDPDEEALRR SRERFETERN SPRFAKLRNW HHGLSAQILN VKS | |
Greater than 80% as determined by SDS-PAGE. |
11155 | |
500 μg | |
Not determined. | |
Full Length of Isoform 6 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |