missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human L Selectin Protein
A cDNA sequence encoding the L Selectin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12521-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
L Selectin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MIFPWKCQST QRDLWNIFKL WGWTMLCCDF LAHHGTDCWT YHYSEKPMNW QRARRFCRDN YTDLVAIQNK AEIEYLEKTL PFSRSYYWIG IRKIGGIWTW VGTNKSLTEE AENWGDGEPN NKKNKEDCVE IYIKRNKDAG KWNDDACHKL KAALCYTASC QPWSCSGHGE CVEIINNYTC NCDVGYYGPQ CQFVIQCEPL EAPELGTMDC THPLGNFSFS SQCAFSCSEG TNLTGIEETT CGPFGNWSSP EPTCQVIQCE PLSAPDLGIM NCSHPLASFS FTSACTFICS EGTELIGKKK TICESSGIWS NPSPICQKLD KSFSMIKEGD YNPLFIPVAV MVTAFSGLAF IIWLARRLKK GKKSKRSMND PY | |
Greater than 95% as determined by SDS-PAGE. Single band on Western Blot. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
L-Sel is supplied in 1x PBS and 50% Glycerol. |