missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KLK7 / PRSS6 Protein

A DNA sequence encoding the Homo sapiens (Human) KLK7 / PRSS6, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6267-ye-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052060

  • 1350.00€ / 500 microgramos


Fecha estimada de envío
30-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001193982.1
KLK7 / PRSS6 Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
5650
Greater than 90% as determined by SDS-PAGE.
Research Use Only
KLK7
Recombinant Protein
Yeast
DKIIDGAPCA RGSHPWQVAL LSGNQLHCGG VLVNERWVLT AAHCKMNEYT VHLGSDTLGD RRAQRIKASK SFRHPGYSTQ THVNDLMLVK LNSQARLSSM VKKVRLPSRC EPPGTTCTVS GWGTTTSPDV TFPSDLMCVD VKLISPQDCT KVYKDLLENS MLCAGIPDSK KNACNGDSGG PLVCRGTLQG LVSWGTFPCG QPNDPGVYTQ VCKFTKWIND TMKKHR
Videos
Documentos