missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human KLF4 His-Cam Protein

Highly purified. Generating reliable and reproducible results.

Marca:  Novus Biologicals™ NBP1-72390-50UG

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214514

  • 516.19€ / 50 microgramos
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

A recombinant protein with a N-Terminal His-Cam tag and corresponding to the amino acids 11-395 of Human KLF4 The Recombinant Human KLF4 Protein is derived from E. coli. The Recombinant Human KLF4 Protein has been validated for the following applications: SDS-Page.
Especificaciones

Especificaciones

NP_004226
Liquid. In 20mM Tris-HCl buffer(pH 8.0) containing 10% glycerol, 1mM DTT, 0.1M NaCl.
58.1kDa
SDS-PAGE
Human
SDS-PAGE
9314
KLF4 Protein
−80°C. Avoid freeze-thaw cycles.
>80%
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales