missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KIR2DL5A Protein

A cDNA sequence encoding the KIR2DL5A was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12490-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15920589

  • 103.00€ / 2 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

KIR2DL5A Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
KIR2DL5A protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
ADPHEGGQDK PLLSAWPSAV VPRGGHVTLL CRSRLGFTIF SLYKEDGVPV PELYNKIFWK SILMGPVTPA HAGTYRCRGS HPRSPIEWSA PSNPLVIVVT GLFGKPSLSA QPGPTVRTGE NVTLSCSSRS SFDMYHLSRE GRAHEPRLPA VPSVNGTFQA DFPLGPATHG GTYTCFGSLH DSPYEWSDPS DPLLVSVTGN SSSSSSSPTE PSSKTGIRRH VEPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVV DVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVS NKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESN GQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHN HYTQKSLSLS PGKHHHHHH
Videos