missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ketohexokinase Protein

A DNA sequence encoding the Homo sapiens (Human) Ketohexokinase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6260-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052000

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000212.1
Ketohexokinase Protein
500 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
3795
Greater than 80% as determined by SDS-PAGE.
Research Use Only
KHK
Recombinant Protein
E. coli
MEEKQILCVG LVVLDVISLV DKYPKEDSEI RCLSQRWQRG GNASNSCTVL SLLGAPCAFM GSMAPGHVAD FVLDDLRRYS VDLRYTVFQT TGSVPIATVI INEASGSRTI LYYDRSLPDV SATDFEKVDL TQFKWIHIEG RNASEQVKML QRIDAHNTRQ PPEQKIRVSV EVEKPREELF QLFGYGDVVF VSKDVAKHLG FQSAEEALRG LYGRVRKGAV LVCAWAEEGA DALGPDGKLL HSDAFPPPRV VDTLGAGDTF NASVIFSLSQ GRSVQEALRF GCQVAGKKCG LQGFDGIV
Videos
Documentos