missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCND2 Protein
A DNA sequence encoding the Homo sapiens (Human) KCND2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9228-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_036413.1 | |
KCND2 Protein | |
Research Use Only | |
KCND2 | |
Recombinant Protein | |
Yeast | |
VSNFSRIYHQ NQRADKRRAQ KKARLARIRA AKSGSANAYM QSKRNGLLSN QLQSSEDEQA FVSKSGSSFE TQHHHLLHCL EKTTNHEFVD EQVFEESCME VATVNRPSSH SPSLSSQQGV TSTCCSRRHK KTFRIPNANV SGSHQGSIQE LSTIQIRCVE RTPLSNSRSS LNAKMEECVK LNCEQPYVTT AIISIPTPPV TTPEGDDRPE SPEYSGGNIV RVSAL | |
Greater than 90% as determined by SDS-PAGE. |
3751 | |
50 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |