missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KAT2A Protein
A cDNA sequence encoding the KAT2A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12477-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
KAT2A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGGGSNSSLS LDSAGAEPMP GEKRTLPENL TLEDAKRLRV MGDIPMELVN EVMLTITDPA AMLGPETSLL SANAARDETA RLEERRGIIE FHVIGNSLTP KANRRVLLWL VGLQNVFSHQ LPRMPKEYIA RLVFDPKHKT LALIKDGRVI GGICFRMFPT QGFTEIVFCA VTSNEQVKGY GTHLMNHLKE YHIKHNILYF LTYADEYAIG YFKKQGFSKD IKVPKSRYLG YIKDYEGATL MECELNPRIP YTELSHIIKK QKEIIKKLIE RKQAQIRKVY PGLSCFKEGV RQIPVESVPG IRETGWKPLG KEKGKELKDP DQLYTTLKNL LAQIKSHPSA WPFMEPVKKS EAPDYYEVIR FPIDLKTMTE RLRSRYYVTR KLFVADLQRV IANCREYNPP DSEYCRCASA LEKFFYFKLK EGGLIDK | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
KAT2A protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 40% glycerol, 200mM NaCl and 1mM EDTA. |