missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IRF 1 Protein

A cDNA sequence encoding the IRF 1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12456-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15910479

  • 160.00€ / 20 microgramos


Fecha estimada de envío
13-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

3659
Greater than 90.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
IRF1
Human
His
1 mg/ml in 20mM Tris pH 8 and 10% glycerol.
IRF 1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MPITRMRMRP WLEMQINSNQ IPGLIWINKE EMIFQIPWKHAAKHGWDINK DACLFRSWAI HTGRYKAGEK EPDPKTWKAN FRCAMNSLPD IEEVKDQSRN KGSSAVRVYR MLPP
Videos