missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Interleukin-12 subunit beta Protein

A DNA sequence encoding the Homo sapiens (Human) Interleukin-12 subunit beta, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8554-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055241

  • 1926.00€ / 1 miligramo


Fecha estimada de envío
22-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_002178.2
Interleukin-12 subunit beta Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
3593
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IL-12
Recombinant Protein
E. coli
IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS
Videos
Documentos