missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL6 / Interleukin-6 Protein

A DNA sequence encoding the Human IL6 / Interleukin-6 Protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10313-ec-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923468

Cantidad Precio
1 267.00€ / 20 microgramos
6 249.00€ / 20 microgramos
Fecha estimada de envío
09-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_000591.1
IL6 / Interleukin-6 Protein
20 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
VPPGEDSKDV AAPHRQPLTS SERIDKQIRY ILDGISALRK ETCNKSNMCE SSKEALAENN LNLPKMAEK DGCFQSGFNE ETCLVKIITG LLEFEVYLEY LQNRFESSEE QARAVQMSTK VLIQFLQKK AKNLDAITTP DPTTNASLLT KLQAQNQWLQ DMTTHLILRS FKEFLQSSLR ALRQM
3569
Greater than 96% as determined by SDS-PAGE and HPLC.
Research Use Only
IL6
Assay 1: The ED50 as determined by a cell proliferation assay using IL-6-dependent murine 7TD1 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg. Assay 2: The ED50 as determined by a cell proliferation assay using IL-6-dependent murine T1165 cells is less than 0.8 ng/ml, corresponding to a specific activity of > 1.25 M 106 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4. , lyophilized
Videos
Documentos