missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL36G / IL1F9 Protein
A DNA sequence encoding the Human IL36G / IL1F9, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10341-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001265497.1 | |
IL36G / IL1F9 Protein | |
Research Use Only | |
IL36G | |
The specific activity is determined by its binding ability in a functional ELISA. Immobilized rHuIL-36gamma at 1 ug/mL can bind recombinant human IL-1 Rrp2 Fc Chimera with a range of 0.15-5 ug/mL. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
56300 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MRGTPGDADG GGRAVYQSMC KPITGTINDL NQQVWTLQGQ NLVAVPRSDS VTPVTVAVIT CKYPEALEQG RGDPIYLGIQ NPEMCLYCEK VGEQPTLQLK EQKIMDLYGQ PEPVKPFLFY RAKTGRTSTL ESVAFPDWFI ASSKRDQPII LTSELGKSYN TAFELNIND | |
Greater than 95% as determined by SDS-PAGE and HPLC. |