missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL36B / IL1F8 Protein

A DNA sequence encoding the Human IL36B / IL1F8, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10339-ec-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913558

  • 114.00€ / 2 microgramos


Fecha estimada de envío
22-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_055253.2
IL36B / IL1F8 Protein
2 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MNPQREAAPK SYAIRDSRQM VWVLSGNSLI AAPLSRSIKP VTLHLIACRD TEFSDKEKGN MVYLGIKGKD LCLFCAEIQG KPTLQLKEKN IMDLYVEKKA QKPFLFFHNK EGSTSVFQSV SYPGWFIATS TTSGQPIFLT KERGITNNTN FYLDSVE
27177
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
Il36b
The specific activity is determined by its binding ability in a functional ELISA. Immobilized rHuIL-36beta at 1 ug/mL can bind recombinant human IL-1 Rrp2 Fc Chimera with a range of 0.15-5 ug/mL.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos