missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL22 / Interleukin 22 Protein

A DNA sequence encoding the Human IL22 / Interleukin 22, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10327-ec-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923518

Cantidad Precio
1 105.00€ / 2 microgramos
10 98.00€ / 2 microgramos
Fecha estimada de envío
04-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_065386.1
IL22 / Interleukin 22 Protein
2 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MAPISSHCRL DKSNFQQPYI TNRTFMLAKE ASLADNNTDV RLIGEKLFHG VSMSERCYLM KQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQKLKDTV KKLGESGEIK AIGELDLLFM SLRNACI
50616
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
IL22
The ED50 as determined by inducing IL-10 secretion of human COLO 205 cells is less than 0.3 ng/ml, corresponding to a specific activity of > 3.3 M 106 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 5.0, lyophilized
Videos
Documentos