missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL20 / Interleukin-20 Protein
Recombinant Protein
Marca: enQuireBio™ QP1165-1 mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_061194.2 | |
IL20 / Interleukin-20 Protein | |
Research Use Only | |
IL20 | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
50604 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MLKTLNLGSC VIATNLQEIR NGFSEIRGSV QAKDGNIDIR ILRRTESLQD TKPANRCCLL RHLLRLYLDR VFKNYQTPDH YTLRKISSLA NSFLTIKKDL RLCHAHMTCH CGEEAMKKYS QILSHFEKLE PQAAVVKALG ELDILLQWME ETE | |
Greater than 95% as determined by SDS-PAGE. |