missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL1F6 / IL36 Protein
A DNA sequence encoding the Human IL1F6 / IL36, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10330-ec-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_055255.1 | |
IL1F6 / IL36 Protein | |
Research Use Only | |
Il36a | |
The specific activity determined by its ability in a functional ELISA. Immobilized rHuIL-36alpha at 1 ug/mL can bind recombinant human IL-1 Rrp2 Fc Chimera with a range of 0.15-5 ug/mL. | |
Human | |
Untagged | |
0.2 um filtered 2 M PBS, pH 7.4, lyophilized |
27179 | |
2 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MEKALKIDTP QQGSIQDINH RVWVLQDQTL IAVPRKDRMS PVTIALISCR HVETLEKDRG NPIYLGLNGL NLCLMCAKVG DQPTLQLKEK DIMDLYNQPE PVKSFLFYHS QSGRNSTFES VAFPGWFIAV SSEGGCPLIL TQELGKANTT DFGLTMLF | |
Greater than 95% as determined by SDS-PAGE and HPLC. |