missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL1F10 / IL-38 Protein
A DNA sequence encoding the Homo sapiens (Human) IL1F10 / IL-38, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7890-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_115945.4 | |
IL1F10 / IL-38 Protein | |
Research Use Only | |
Il1f10 | |
Recombinant Protein | |
E. coli | |
MCSLPMARYY IIKYADQKAL YTRDGQLLVG DPVADNCCAE KICILPNRGL ARTKVPIFLG IQGGSRCLAC VETEEGPSLQ LEDVNIEELY KGGEEATRFT FFQSSSGSAF RLEAAAWPGW FLCGPAEPQQ PVQLTKESEP SARTKFYFEQ SW | |
Greater than 90% as determined by SDS-PAGE. |
84639 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |