missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL17F / IL-17F Protein

A DNA sequence encoding the Human IL17F / IL-17F, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10322-ec-25ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913498

  • 299.00€ / 25 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

NP_443104.1
IL17F / IL-17F Protein
25 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MRKIPKVGHT FFQKPESCPP VPGGSMKLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGKEDISMN SVPIQQETLV VRRKHQGCSV SFQLEKVLVT VGCTCVTPVI HHVQ
112744
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
IL17F
The ED50 as determined by inducing IL-6 secretion of murine NIH/3T3 cells is less than 20 ng/ml, corresponding to a specific activity of > 5.0 M 104 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos