missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-9 / Interleukin-9 Protein

A DNA sequence encoding the Human IL-9 / Interleukin-9, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10317-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933478

Cantidad Precio
1 1910.00€ / 100 microgramos
3 1782.03€ / 100 microgramos
Fecha estimada de envío
28-10-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_000581.1
IL-9 / Interleukin-9 Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
QGCPTLAGIL DINFLINKMQ EDPASKCHCS ANVTSCLCLG IPSDNCTRPC FSERLSQMTN TTMQTRYPLI FSRVKKSVEV LKNNKCPYFS CEQPCNQTTA GNALTFLKSL LEIFQKEKMR GMRGKI
3578
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
IL9
The ED50 as determined by a cell proliferation assay using human MO7e cells is less than 0.2 ng/ml, corresponding to a specific activity of > 5.0 M 106 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos